Order Entry
Puerto Rico
ContactUsLinkComponent
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit, AAT Bioquest
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit, AAT Bioquest
Catalog # 76484-832
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit, AAT Bioquest
Catalog # 76484-832
Supplier:  AAT Bioquest
Supplier Number:  22415
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    MycoLight™ Rapid fluorescence gram-positive bacteria staining kit
  • Size:
    100 Tests
  • Cat. No.:
    76484-832
  • No. of tests:
    100

Specifications

About this item

MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit provides a novel one-step fluorescence assay for the determination of gram sign in living bacteria.

  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for fluorescent microscopy
Caution: For research use only.