Order Entry
Puerto Rico
ContactUsLinkComponent
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*, AAT Bioquest
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*, AAT Bioquest
Catalog # 76484-498
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*, AAT Bioquest
Catalog # 76484-498
Supplier:  AAT Bioquest
Supplier Number:  22401
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    MycoLight™ Ratiometric bacterial membrane potential kit *red/green fluorescence*
  • Size:
    200 Tests
  • Cat. No.:
    76484-498
  • No. of tests:
    200

Specifications

About this item

AAT Bioquest's MycoLight™ Ratiometric Bacterial Membrane Potential Kit uses a fluorescent sensor that exhibits green fluorescence in both gram-positive and negative bacterial cells in low concentration, but the fluorescence shifts toward red emission at higher cytosolic concentrations due to the dye molecule aggregation caused by larger membrane potentials.

  • Suitable for gram-positive and gram-negative bacteria
  • Ratiometric detection
  • Contains all necessary Components
  • Multiple applications
Caution: For research use only.