Cell Meter™ Caspase 9 Activity Apoptosis Assay Kit *Blue Fluorescence*, AAT Bioquest
Catalog # 76483-770
Supplier: AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ Caspase 9 activity apoptosis assay kit *blue fluorescence*
- Size:200 Tests
- Cat. No.:76483-770
- No. of tests:200
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cellular functions.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.