PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Enhanced Selectivity, AAT Bioquest
76483-440
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Description:PhosphoWorks™ Fluorimetric pyrophosphate assay kit *enhanced selectivity*
- Size:200 Tests
- Cat. No.:76483-440
- No. of tests:200
Pyrophosphate (PPi) is produced by a number of biochemical reactions, such as ATP hydrolysis, DNA and RNA polymerizations, cyclic AMP formation by the enzyme adenylate cyclase and the enzymatic activation of fatty acids to form their coenzyme A esters.
- Pyrophosphate-dependent fluorescence
- Highly selective for PPi
- Contains all necessary components
: For research use only.