ContactUsLinkComponent
Cell Meter™ Flow Cytometric Calcium Assay Kit, AAT Bioquest
  76484-684
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
undefined
Cell Meter™ Flow Cytometric Calcium Assay Kit, AAT Bioquest
  76484-684
 :  36310
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Cell Meter™ Flow cytometric calcium assay kit
  • Size:
    100 Tests
  • Cat. No.:
    76484-684
  • No. of tests:
    100
Cell Meter™ Flow Cytometric Calcium assay kit provides fluorescence-based assays for detecting intracellular calcium mobilization using a flow cytometer.
  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimized for flow cytometers but suitable for all major fluorescence platforms
  • Multiple research applications
 : For research use only.