Screen Quest™ Fluo-8 Medium Removal Calcium Assay Kit, Optimized for Difficult Cell Lines, AAT Bioquest
Supplier: AAT BIOQUEST INC
Certificates
About this item
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
- 100 to 250 times fluorescence increase once bound to calcium
- Contains all necessary components
- Suitable for multiple types of cells
- Robust and adaptable protocol
- Brighter and more photostable signal
Caution: For research use only.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Related Information
Manual Fluo-8 Medium Removal Calcium Assay Kit, Optimized for Difficult Cell Line.pdf Manual Screen Quest Fluo 8 Medium Removal Calcium Assay Kit Optimized for Difficult Cell Lines.pdf Manual Screen Quest™ Fluo 8 Medium Removal Calcium Assay Kit Optimized For Difficult Cell Lines.pdf Manual Screen Quest™ Fluo-8 Medium Removal Calcium Assay Kit Optimized for Difficult Cell Lines.pdf