Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Orange Fluorescence Optimized for Flow Cytometry*, AAT Bioquest
Catalog # 76483-760
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ Mitochondrion membrane potential assay kit *Orange fluorescence optimized for flow cytometry*
- Size:100 Tests
- Cat. No.:76483-760
- No. of tests:100
Specifications
About this item
This particular kit is designed to detect cell apoptosis by measuring the loss of the mitochondrial membrane potential(MMP).
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.