Live or Dead™ Cell Viability Assay Kit *Green/Red Dual Fluorescence*, AAT Bioquest
This Live or Dead™cell viability uses two fluorogenic indicators: calcein AM for viable cells and a cell-impermeable DNA-binding dye for the cells with compromised membranes.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
- Results in under 2 hours
: For research use only.
Frequently Bought Together