Specifications
- Description:MycoLight™ Fluorescence live/dead bacterial Imaging kit
- Size:100 Tests
- Cat. No.:76484-822
- No. of tests:100
Specifications
About this item
AAT Bioquest's MycoLight™ Fluorescence Live/Dead Bacterial Imaging Kit provides two-color fluorescence assay for visualizing live and dead bacteria through fluorescent microscope.
- Suitable for gram-positive and gram-negative bacteria
- Contains all necessary components
- Multiple applications
- Optimized for fluorescent microscopy