MycoLight™ Fluorimetric CTC Live Bacteria Quantification Kit, AAT Bioquest
76484-738
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Description:MycoLight™ Fluorimetric CTC live bacteria quantification kit
- Size:100 Tests
- Cat. No.:76484-738
- No. of tests:100
The CTC Flow Cytometric live bacteria assay kit provides an easy and convenient method for evaluating bacterial health and vitality as a function of the respiratory activity.
- Suitable for gram-positive and gram-negative bacteria
- Contains all necessary components
- Multiple applications
- Optimized for flow cytometry
: For research use only.