ContactUsLinkComponent
MESG *Phosphate Assay Reagent*, AAT Bioquest
MESG *Phosphate Assay Reagent*, AAT Bioquest
Catalog # 76483-424
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
MESG *Phosphate Assay Reagent*, AAT Bioquest
Catalog # 76483-424
Supplier:  AAT Bioquest
Supplier Number:  21600
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    MESG *Phosphate assay reagent*
  • Size:
    5 mg
  • Cat. No.:
    76483-424
Specifications
About this item
In the presence of inorganic phosphate MESG is converted to 2-amino-6-mercapto-7-methlpurine by purine nucleoside phosphorylase (EC 2.4.2.1) with absorption wavelength shift to red.
  • Adaptable and robust
  • Quantifiable emission wavelength shift
  • Standalone reagent
Caution: For research use only.