Order Entry
Puerto Rico
ContactUsLinkComponent
Native Human Concanavalin A (from Human), iFluor® 555
Native Human Concanavalin A (from Human), iFluor® 555
Catalog # 76485-370
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Native Human Concanavalin A (from Human), iFluor® 555
Catalog # 76485-370
Supplier:  AAT BIOQUEST INC
Supplier Number:  25585
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Conjugation:
    iFluor® 555
  • Protein/Peptide Type:
    Native
  • Species:
    Human
  • Size:
    1 mg
  • Protein Synonyms:
    ConA
  • Protein/Peptide Name:
    Concanavalin A
  • Cat. No.:
    76485-370

Specifications

About this item

Concanavalin A (ConA) is a lectin that binds specifically to certain structures found in various sugars, glycoproteins and glycolipids.

  • Well-cited conjugate
  • High affinity and selectivity
  • Multiple applications
Caution: For research use only.