ReadiUse™ Disposable PD-10 Desalting Column, AAT Bioquest
Supplier: AAT BIOQUEST INC
Certificates
About this item
PD-10 Desalting Columns contain Sephadex G-25 resin for rapid buffer exchange, desalting, and removal of small contaminants (salts, dyes, radioactive labels) from samples using gravity flow or centrifugation.
- Efficient contaminant removal
- Excellent sample recovery
- Dialysis alternative
Caution: For research use only.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.