Cell Meter™ Caspase 9 Activity Apoptosis Assay Kit *Red Fluorescence*, AAT Bioquest
Catalog # 76483-774
Supplier: AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ caspase 9 activity apoptosis assay kit *red fluorescence*
- Size:100 Tests
- Cat. No.:76483-774
- No. of tests:100
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cellular functions.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.