Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Orange Fluorescence Optimized for Microplate Reader*, AAT Bioquest
Catalog # 76483-762
Supplier: AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ mitochondrion membrane potential assay kit *orange fluorescence optimized for microplate reader*
- Size:500 Tests
- Cat. No.:76483-762
- No. of tests:500
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cellular functions.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for microplate readers but suitable for all major fluorescence platforms
Caution: For research use only.