Order Entry
Puerto Rico
ContactUsLinkComponent
Live or Dead™ Fixable Dead Cell Staining Kit *Blue Fluorescence*, AAT Bioquest
Live or Dead™ Fixable Dead Cell Staining Kit *Blue Fluorescence*, AAT Bioquest
Catalog # 76483-608
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Live or Dead™ Fixable Dead Cell Staining Kit *Blue Fluorescence*, AAT Bioquest
Catalog # 76483-608
Supplier:  AAT BIOQUEST INC
Supplier Number:  22600
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Live or Dead™ fixable dead cell staining kit *blue fluorescence*
  • Size:
    200 Tests
  • Cat. No.:
    76483-608
  • No. of tests:
    200

Specifications

About this item

Our Live or Dead™ Fixable Dead Cell Staining Kits are a set of tools for labeling cells for fluorescence microscopic investigations of cell functions.

  • Contains all necessary components
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.