ReadiUse™ NADP Regenerating Kit, AAT Bioquest
Catalog # 76482-358
Supplier: AAT Bioquest
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:ReadiUse™ NADP regenerating kit
- Size:200 tests
- Cat. No.:76482-358
- No. of tests:200
Specifications
About this item
NADP is the electron acceptor for biosynthetic reactions and dehydrogenation reactions.
- Ready to use
- Robust protocol
- Contains all necessary components
Caution: For research use only.