Order Entry
Puerto Rico
ContactUsLinkComponent
ReadiUse™ TCA Deproteinization Sample Preparation Kit, AAT Bioquest
ReadiUse™ TCA Deproteinization Sample Preparation Kit, AAT Bioquest
Catalog # 76482-958
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ReadiUse™ TCA Deproteinization Sample Preparation Kit, AAT Bioquest
Catalog # 76482-958
Supplier:  AAT BIOQUEST INC
Supplier Number:  19501
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ReadiUse™ TCA deproteinization sample preparation kit
  • Size:
    200 tests
  • Cat. No.:
    76482-958
  • No. of tests:
    200

Specifications

About this item

The analysis of amino acids, ions, metabolites and other small molecules is frequently hindered by the interference of lipids, protein and enzymes present in biofluids, cell and tissue lysates.

  • Contains all necessary components
  • Useful for preparing small molecules
  • Can be used in small- and large-scale
Caution: For research use only.