StrandBrite™ Green Fluorimetric RNA Quantitation Kit *High Selectivity*, AAT Bioquest
Catalog # 76482-952
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:StrandBrite™ green fluorimetric RNA quantitation kit *high selectivity*
- Size:100 tests
- Cat. No.:76482-952
- No. of tests:100
Specifications
About this item
The major challenge to analyze RNA in live cells is the interferences caused by DNA.
- Highly sensitive RNA detection
- Robust protocol
- Multiple applications
Caution: For research use only.