Order Entry
Puerto Rico
ContactUsLinkComponent
ReadiUse™ dNTP Mix *10 mM* (5 ml), AAT Bioquest
ReadiUse™ dNTP Mix *10 mM* (5 ml), AAT Bioquest
Catalog # 76485-404
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
ReadiUse™ dNTP Mix *10 mM* (5 ml), AAT Bioquest
Catalog # 76485-404
Supplier:  AAT Bioquest
Supplier Number:  17200
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    ReadiUse™ dNTP Mix *10 mm*
  • Size:
    5 ml
  • Cat. No.:
    76485-404

Specifications

About this item

Our Ready-to Use Solution of deoxynucleotide triphosphates (dNTPs) have been extensively tested and verified for use in a wide variety of molecular biology applications, including PCR, real-time PCR, high-fidelity PCR, long PCR, cDNA synthesis, RT-PCR, RCA, MDA, DNA labeling, DNA sequencing, in vitro transcription, siRNA synthesis, and RNA amplification.

  • Ready to use formulation
  • In-house testing and quality control
  • Multiple applications
Caution: For research use only.