Orum International VERI.GAS™ Microbial Compressed Air/Gas Sampler
Catalog # 76509-314
Supplier: Hardy Diagnostics
Specifications
- Description:VERI.GAS microbial compressed air/gas sampler
- Cat. No.:76509-314
Specifications
About this item
The VERI.GAS is an accessory for compressed air and airflow rate checks. Test for the presence of microorganisms in compressed air/gas and verify the calibrated air flow rate is within appropriate specification.
- Suitable for testing compressed air and nitrogen
- Fully autoclavable
- Compatible with 100Lpm air sampling instruments (TRIO.BAS, AIRBIO, and MULTIFLEX models)
- Includes a manual verification instrument to check the precision of airflow rate calibration
Includes one digital control unit, one stainless steel bell chamber for gas testing, one technopolymer bell chamber for calibration specification check, one connection tube, and a carrying case.
Accessories information: AIRBIO ONE not included.