Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant CD45/PTPRC Protein (aa 453-1152, His & GST Tag) (from Baculovirus Insect Cells)
 
undefined
Mouse Recombinant CD45/PTPRC Protein (aa 453-1152, His & GST Tag) (from Baculovirus Insect Cells)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    1. Measured by its ability to cleave a substrate, pNitrophenyl phosphate (pNPP). The specific activity is >50,000 pmoles/min/μg.
    2. Measured by its ability to bind biotinylated Galectin-1 (Cat:10290-HNAE-E) in a functional ELISA.
  • Gene ID:
    AAA39458.1
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    CD45R|B220|CD45|GP180|T200|L-CA|LY5|LCA
  • Protein/Peptide Name:
    PTPRC
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse PTPRC/GST chimera consists of 937 amino acids and has a calculated molecular mass of 108.2 kDa. The recombinant protein migrates as an approximately 110 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Arg453-Ser1152
  • Shipping Temperature:
    Ambient