SARS-CoV-2 (2019-nCoV) Recombinant Helicase-His Protein (from E. coli)
Catalog # 103886-012
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Gene ID:YP_009725308.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:HE-CoV
- Protein/Peptide Name:HE-CoV
- Purity:>80% as determined by SDS-PAGE.
- Molecular Weight:The recombinant SARS-CoV-2 (2019-nCoV) Helicase-His Recombinant Protein consists of 608 amino acids and predicts a molecular mass of 67.8 kDa.
- Sequence:Ala1-Gln601
- Shipping Temperature:Ambient
- Cat. No.:103886-012
Specifications
About this item
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Helicase-His Recombinant Protein (YP_009725308.1) (Ala1-Gln601) was expressed with a polyhistidine tag at the N-terminus.
- Shipping temperature: Dry ice