Order Entry
Puerto Rico
ContactUsLinkComponent
Influenza A H5N1 (A/common magpie/Hong Kong/5052/2007) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Influenza A H5N1 (A/common magpie/Hong Kong/5052/2007) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Catalog # 103663-574
Supplier:  Sino Biological
CAS Number:  
undefined
Influenza A H5N1 (A/common magpie/Hong Kong/5052/2007) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Catalog # 103663-574
Supplier:  Sino Biological
Supplier Number:  40044-V08B-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    H5N1
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Biological Activity:
    Measured by its ability to agglutinate guinea pig red blood cells.
    HA titer is 0.02-0.1 μg/ml for 1%GRBC.
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Hemagglutinin
  • Protein/Peptide Name:
    HA
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant hemagglutinin of Influenza A virus (A/common magpie/Hong Kong/5052/2007(H5N1)) consists 524 amino acids and predicts a molecular mass of 59.6 kDa.
  • Sequence:
    Met1-Gln530
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103663-574

Specifications

About this item

A DNA sequence encoding the Influenza A virus (A/common magpie/Hong Kong/5052/2007(H5N1)) hemagglutinin (Met1-Gln530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.