Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant MSRB3 Protein (His Tag) (from E. coli)
 
undefined
Human Recombinant MSRB3 Protein (His Tag) (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    Please contact us for more information.
  • Gene ID:
    Q8IXL7-2
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    DFNB74
  • Protein/Peptide Name:
    MSRB3
  • Purity:
    >95% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human MSRB3 consists of 178 amino acids and predicts a molecular mass of 19.6 KDa. It migrates as an approximately 23 KDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Gly26-Leu185
  • Shipping Temperature:
    Ambient