Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant B7-H5/Gi24/VISTA Protein (ECD, Fc Tag) (from HEK293 Cells)
  103623-078
 :  
undefined
Human Recombinant B7-H5/Gi24/VISTA Protein (ECD, Fc Tag) (from HEK293 Cells)
  103623-078
 :  13482-H02H-100
 :  

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to bind human IGSF11-His in a functional ELISA.
  • Gene ID:
    AAH20568.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    B7-H5|C10orf54|GI24|PP2135|DD1alpha|SISP1|B7H5|VISTA|PD-1H
  • Protein/Peptide Name:
    Vsir
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant human C10orf54 consists 400 amino acids and predicts a molecular mass of 44.9 kDa.
  • Sequence:
    Met1-Ala194
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103623-078

 

 

A DNA sequence encoding the human C10orf54 (AAH20568.1) (Met1-Ala194) was expressed with the Fc region of human IgG1 at the C-terminus.