Human Recombinant TSC22D1 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human TSC22D1 isoform 2 (Q15714-2) (Met 1-Ala 144) was expressed, with a polyhistidine tag at the N-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information
- Gene ID:Q15714-2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TSC22|TGFB1I4|Ptg-2
- Protein/Peptide Name:TSC22D1
- Purity:>92% as determined by SDS-PAGE
- Molecular Weight:The recombinant human TSC22D1 consisting of 155 amino acids and has a calculated molecular mass of 17.2 kDa. It migrates as an 20 kDa band in SDS-PAGE under reducing conditions as predicted.
- Sequence:Met1-Ala144
- Shipping Temperature:Ambient