Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant STXBP1/UNC18A Protein (His & GST Tag) (from Baculovirus Insect cells)
Human Recombinant STXBP1/UNC18A Protein (His & GST Tag) (from Baculovirus Insect cells)
Supplier:  Sino Biological
Certificates
undefined
Human Recombinant STXBP1/UNC18A Protein (His & GST Tag) (from Baculovirus Insect cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Gene ID:
    P61764-1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    MUNC18-1|P67|RBSEC1|UNC18|NSEC1
  • Protein/Peptide Name:
    STXBP1
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human STXBP1/GST chimera consists of 831 amino acids and has a calculated molecular mass of 95.4 kDa. It migrates as an approximately 80 kDa band in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Ser594
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together