Influenza A H5N2 (A/American green-winged teal/California/HKWF609/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from HEK293 Cells)
Catalog # 103622-324
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Influenza A
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:ACF47563.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>96% as determined by SDS-PAGE
- Molecular Weight:The secreted recombinant influenza A H5N2 HA (A/American green-winged teal/California/HKWF609/07 (H5N2)) comprises 522 amino acids and has a predicted molecular mass of 59.5 kDa. As a result of glycosylation, it migrates as an approximately 60-70 kDa band in SDS-PAGE under reducing conditions.
- Sequence:Met1-Gln527
- Shipping Temperature:Ambient
- Cat. No.:103622-324
Specifications
About this item
A DNA sequence encoding the extracellular domain (Met 1-Gln 527) of the influenza A hemagglutinin (A/American green-winged teal/California/HKWF609/2007(H5N2)) (ACF47563.1) (HA1+HA2, uncleaved), was expressed, fused with a C-terminal polyhistidine tag.