Influenza A H1N1 (A/Brisbane/59/2007) Recombinant Hemagglutinin Protein (HA1 Subunit) (His Tag) (from HEK293 Cells)
Catalog # 103632-674
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:H1N1
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:ACA28844.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>98% as determined by SDS-PAGE
- Molecular Weight:Recombinant HA1 subunit of Influenza A virus (A/Brisbane/59/2007 (H1N1)) hemagglutinin consists of 337 amino acids and has a calculated molecular mass of 37.7 kDa. As a result of glycosylation, the recombinant protein migrates as an approximately 55-60 kDa protein in SDS-PAGE under reducing conditions.
- Sequence:Met1-Arg343
- Shipping Temperature:Ambient
- Cat. No.:103632-674
Specifications
About this item
A DNA sequence encoding an N-terminal segment (Met 1-Arg 343) of Influenza A virus (A/Brisbane/59/2007 (H1N1)) (ACA28844.1) HA, termed as HA1 subunit, was fused with a polyhistidine tag at the C-terminus.