Human Recombinant C2/Complement Component 2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human complement component 2 (C2) precursor (NP_000054.2) (Met 1-Leu 752) was expressed with a C-terminal polyhistidine tag.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by its ability to cleave a colorimetric peptide substrate, N-carbobenzyloxy-Gly-Arg-ThioBenzyl ester (Z-GR-SBzl), in the presence of 5,5’Dithiobis (2-nitrobenzoic acid) (DTNB). The specific activity is >100 pmoles/min/μg.
- Gene ID:NP_000054.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CO2|ARMD14
- Protein/Peptide Name:C2
- Purity:>97% as determined by SDS-PAGE.
- Molecular Weight:The single-chain form of recombinant human complement component C2 comprises 743 amino acids and has a calculated molecular mass of 82.5 kDa.
- Sequence:Met1-Leu752
- Shipping Temperature:Ambient