Fetal bovine serum, 50 ml, Premium, United States Origin, Corning®
76418-024
: Corning
- Sterility:Sterile
- Description:Fetal bovine serum
- Size:50 ml
- Origin:United States
- Storage Temperature:−40...−10 °C
- Cat. No.:76418-024
Fetal bovine serum, United States origin collected at government approved abattoirs in the United States.
- Shelf life: 60 months
- Weight: 0.09 kg
- Format: Liquid
True pooled and triple 0.1 µm sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.
Granted a Certificate of Suitability, R0-CEP 2018-271, by the European Directorate for the Quality of Medicines (EDQM).