Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant SIRP alpha/CD172a Protein (Fc Tag) (from HEK293 Cells)
Mouse Recombinant SIRP alpha/CD172a Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103665-622
Supplier:  Sino Biological
CAS Number:  
undefined
Mouse Recombinant SIRP alpha/CD172a Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103665-622
Supplier:  Sino Biological
Supplier Number:  50956-M02H-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its binding ability in a functional ELISA.
    Immobilized rat CD47-His (Cat:80305-R08H) at 10 μg/ml (100 μl/well) can bind mouse SIRPA-Fc, The EC50 of mouse SIRPA-Fc is 0.6-1.4 μg/ml.
  • Gene ID:
    BAA20376.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    PTPNS1|CD172A|MYD-1|SHPS1|P84|SIRP alpha|BIT|SIRP|MFR
  • Protein/Peptide Name:
    SIRPA
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    The secreted recombinant mouse SIRPA/Fc is a disulfide-linked homodimer. The reduced monomer comprises 583 amino acids and has a calculated molecular mass of 64.9 kDa. As a result of glycosylation, the apparent molecular mass of rmSIRPA/Fc monomer is approximately 1001-20 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Asn373
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103665-622

Specifications

About this item

A DNA sequence encoding the extracellular domain of mouse SIRPA (BAA20376.1) (Met 1-Asn 373) was fused with the Fc region of human IgG1 at the C-terminus.