Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant IGF1R/CD221 Protein (His & GST Tag) (from Baculovirus Insect cells)
Catalog # 103623-892
Supplier:  Sino Biological
CAS Number:  
undefined
Human Recombinant IGF1R/CD221 Protein (His & GST Tag) (from Baculovirus Insect cells)
Catalog # 103623-892
Supplier:  Sino Biological
Supplier Number:  10164-H20B1-50
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Insect cells)
  • Species:
    Human
  • Size:
    50 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    The specific activity was determined to be >100 nmol/min/mg using Poly(Glu,Tyr) 4:1 as substrate.
  • Gene ID:
    NP_000866.1
  • Reconstitution Instructions:
    A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    JTK13|CD221|IGFR|IGFIR|IGF1 Receptor|IGF-I R
  • Protein/Peptide Name:
    IGF1R
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human IGF1R (aa 954-1367) /GST chimera consists of 651 amino acids and has a calculated molecular mass of 74.6 KDa.
  • Sequence:
    Met954-Cys1367
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103623-892

Specifications

About this item

A DNA sequence encoding the human IGF1R (NP_000866.1) cytoplasmic domain (Met 954-Cys 1367) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.

  • Shipping temperature: Dry ice