Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant HER2/ErbB2 Protein (ECD, domain IV) (His Tag) (from HEK293 Cells)
Human Recombinant HER2/ErbB2 Protein (ECD, domain IV) (His Tag) (from HEK293 Cells)
Catalog # 103620-670
Supplier:  Sino Biological
CAS Number:  
undefined
Human Recombinant HER2/ErbB2 Protein (ECD, domain IV) (His Tag) (from HEK293 Cells)
Catalog # 103620-670
Supplier:  Sino Biological
Supplier Number:  10004-H08H4-50
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    50 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg protein as determined by the LAL method.
  • Biological Activity:
    Immobilized Her2/ERBB2 Protein, Human, Recombinant (ECD, domain IV, His Tag)(Cat:10004-H08H4) at 2 μg/ml (100 μl/well) can bind Anti-Erbb2 Antibody (Trastuzumab)(Cat:68046-H002), the EC50 is 15-70 ng/ml.
  • Gene ID:
    NP_004439.2
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    NEU|MLN19|HERVW|TKR1|HERV7Q|MLN 19|CD340|ERVWE1|HER2|HERV-W-ENV|ENV|HER-2/neu|HER-2|ENVW|HERVWENV|HERV-7q|NGL
  • Protein/Peptide Name:
    ERBB2
  • Purity:
    >85% as determined by SDS-PAGE
  • Molecular Weight:
    The recombinant human ERBB2 consists of 153 amino acids and predicts a molecular mass of 17.1 kDa. As a result of glycosylation, human ErbB2 migrates as an approximately 27 kDa protein in SDS-PAGE under reducing conditions.
  • Sequence:
    Pro489-Cys630
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103620-670

Specifications

About this item

A DNA sequence encoding the human ERBB2 (NP_004439.2) (Pro489-Cys630) was expressed with a polyhistidine tag at the C-terminus.