Corning® 384-Well Microplate Black/Clear, Round Bottom, Ultra-Low Attachment, Spheroid, Corning
75838-886
: Corning
- No. of wells:384
- Sterility:Sterile
- Well volume:90 µl
- Color:Black/clear
- Description:Round bottom, 384 well
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:RECs, ISO 14001 - Kennebunk
- Cat. No.:75838-886
- Packaging:10/pack, 50/case
- Surface treatment:Ultra-low attachment
- Working well volume:20 - 80 µl
- Cell growth area:0.06 cm²
- Well bottom shape:Round
- Supplier no.:3830BC
Ultra-Low Attachment microplates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment.
- Sterilized by gamma irradiation
- Opaque walls to prevent well-to-well crosstalk
- Surface is noncytotoxic, biologically inert and nondegradable
- With Generic Bar codes, and lid
Surface minimizes protein absorption, enzyme activation and cellular activation; Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk.
Can be used for both top and bottom reading instruments. Novel well geometry aids formation of spheroids in center of well. Optically clear round bottom with black opaque microplate body