Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant Carboxypeptidase A2/CPA2 Protein (His Tag) (from HEK293 Cells)
Mouse Recombinant Carboxypeptidase A2/CPA2 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant Carboxypeptidase A2/CPA2 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to cleave the colorimetric peptide substrate Ac-Phe-Thiaphe-OH in the presence of DTNB.
    The specific activity is >4000 pmoles/min/μg.
  • Gene ID:
    Q504N0
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    Carboxypeptidase A2|CPA2
  • Protein/Peptide Name:
    CPA2
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular Weight:
    The secreted recombinant mouse CPA2 (pro form) comprises 412 amino acids and has a calculated molecular mass of 46.6 kDa. It migrates as an approximately 47 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Met1-Tyr417
  • Shipping Temperature:
    Ambient

Specifications

Related Information