Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant ST6GAL1/CD75 Protein (His Tag) (from HEK293 Cells)
Mouse Recombinant ST6GAL1/CD75 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Mouse Recombinant ST6GAL1/CD75 Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to transfer Neu5Ac from CMP-Neu5Ac to N-Acetyllactosamine.
    The specific activity is > 150 pmol/min/μg.
  • Gene ID:
    NP_666045.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    St6gal|Siat1|St6galI|St6Gal-I|AW742324
  • Protein/Peptide Name:
    ST6GAL1
  • Purity:
    >96% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant mouse ST6GAL1 comprises 396 amino acids and predicts a molecular mass of 45.9 kDa. The apparent molecular mass of mouse ST6GAL1 is approximately 50-55 kDa in SDS-PAGE under reducing conditions.
  • Sequence:
    Lys27-Cys403
  • Shipping Temperature:
    Ambient

Specifications

Related Information