Mouse Recombinant Smad5 Protein (from Baculovirus Insect Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mouse SMAD5 (P97454) (Thr2-Ser465) was expressed and purified with two additional amino acids (Gly and Pro ) at the N-terminus.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokines and Growth Factors
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:P97454
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:AI451355|Dwf-C|Madh5|MusMLP|1110051M15Rik
- Protein/Peptide Name:Smad5
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse SMAD5 consists of 466 amino acids and predicts a molecular mass of 52.2 KDa. It migrates as an approximately 57 KDa band in SDS-PAGE under reducing conditions.
- Sequence:Thr2-Ser465
- Shipping Temperature:Ambient