Mouse Recombinant CXADR Protein (His & Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mouse CXADR (NP_001020363.1) extracellular domain (Met 1-Gly 237) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:Measured by the ability of the immobilized protein to support the adhesion of mouse neutrophils. When 5 x 10E4 cells/well are added to CXADR-coated plates (4 μg/ml and 100 μl/well), approximately 20%-40% will adhere specifically after 60 minutes at 37 ℃.
- Gene ID:NP_001020363.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:CAR4/6|CXADR|HCAR|CAR
- Protein/Peptide Name:CXADR
- Purity:>92% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse CXADR/Fc is a disulfide-linked homodimer. The reduced monomer consists of 466 amino acids and has a predicted molecular mass of 52 kDa. As a result of glycosylation, the apparent molecular mass of rm CXADR/Fc monomer migrates with an apparent molecular mass of 60-65 kDa in SDS-PAGE under reducing conditions.
- Sequence:Met1-Gly237
- Shipping Temperature:Ambient