Human Recombinant LRRC32 Protein (His Tag) (from HEK293 Cells)
Catalog # 103905-994
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_005503.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:GARP|D11S833E
- Protein/Peptide Name:LRRC32
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant human LRRC32 consists of 619 amino acids and predicts a molecular mass of 67.5 kDa.
- Sequence:Met1-Asn627
- Shipping Temperature:Ambient
- Cat. No.:103905-994
Specifications
About this item
A DNA sequence encoding the human LRRC32 (NP_005503.1) (Met1-Asn627) was expressed with a polyhistidine tag at the C-terminus.