Influenza A H3N2 (A/Brisbane/10/2007) Recombinant Hemagglutinin/HA Protein (His Tag) (from Baculovirus Insect cells)
Catalog # 103621-820
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:H3N2
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Gene ID:ABW23353.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant hemagglutinin of Influenza A virus (A/Brisbane/10/2007(H3N2)) consists 525 amino acids and predicts a molecular mass of 59.2 kDa.
- Sequence:Met1-Trp530
- Shipping Temperature:Ambient
- Cat. No.:103621-820
Specifications
About this item
A DNA sequence encoding the Influenza A virus (A/Brisbane/10/2007(H3N2)) hemagglutinin (ABW23353.1) (Met1-Trp530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.