Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Iduronate 2-Sulfatase/IDS Protein (His Tag) (from HEK293 Cells)
Human Recombinant Iduronate 2-Sulfatase/IDS Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological
Certificates
undefined
Human Recombinant Iduronate 2-Sulfatase/IDS Protein (His Tag) (from HEK293 Cells)
Supplier:  Sino Biological

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Biological Activity:
    Measured by its ability to hydrolyze the substrate 4-Nitrocatechol Sulfate (PNCS).
    The specific activity is > 1.0 pmoles/min/μg.
  • Gene ID:
    NP_000193.1
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    MPS2|IDS|SIDS
  • Protein/Peptide Name:
    IDS
  • Purity:
    >87% as determined by SDS-PAGE
  • Molecular Weight:
    The secreted recombinant human IDS with the propeptide consists of 536 amino acids after removal of the signal peptide and has a predicted molecular mass of 61 kDa. In SDS-PAGE under reducing conditions, it migrates as several bands with apparent molecular mass between 85-95 KDa due to glycosylation.
  • Sequence:
    Met1-Pro550
  • Shipping Temperature:
    Ambient

Specifications

Related Information