Rhesus Recombinant B7-H5/GI24/VISTA Protein (ECD, His Tag) (from HEK293 Cells)
103620-826
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:NP_001177807.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:B7-H5|C10orf54|GI24|PP2135|DD1alpha|SISP1|B7H5|VISTA|PD-1H
- Protein/Peptide Name:Vsir
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rhesus C10orf54 consists 173 amino acids and predicts a molecular mass of 19.6 kDa.
- Sequence:Met1-Ala194
- Shipping Temperature:Ambient
- Cat. No.:103620-826
A DNA sequence encoding the rhesus C10orf54 (NP_001177807.1) (Met1-Ala194) was expressed with a polyhistidine tag at the C-terminus.