Order Entry
Puerto Rico
ContactUsLinkComponent
Cynomolgus Recombinant EDAR Protein (His Tag) (from HEK293 Cells)
Cynomolgus Recombinant EDAR Protein (His Tag) (from HEK293 Cells)
Catalog # 103585-376
Supplier:  Sino Biological
CAS Number:  
Cynomolgus Recombinant EDAR Protein (His Tag) (from HEK293 Cells)
Catalog # 103585-376
Supplier:  Sino Biological
Supplier Number:  90111-C08H-100
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Immunotherapy
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Size:
    100 µg
  • Storage Conditions:
    –20…–80 °C
  • Endotoxin Content:
    <1.0 EU per μg of the protein as determined by the LAL method
  • Gene ID:
    F7GZI3
  • Reconstitution Instructions:
    A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    ECTD10B|ED5|ED3|ED1R|HRM1|EDA-A1R|ECTD10A|EDA1R|EDA3|DL
  • Protein/Peptide Name:
    EDAR
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    The recombinant cynomolgus EDAR comprises 172 amino acids and has a calculated molecular mass of 18.8 KDa. The apparent molecular mass of it is approximately 30-40 KDa respectively in SDS-PAGE.
  • Sequence:
    Met1-Ala187
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    103585-376

Specifications

About this item

A DNA sequence encoding the cynomolgus EDAR (F7GZI3) (Met1-Ala187) was expressed with a polyhistidine tag at the C-terminus.