Rhesus Recombinant SFTPD/SP-D/SFTP4 Protein (His Tag) (from HEK293 Cells)
103670-100
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Rhesus monkey
- Size:50 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Gene ID:NP_001035283.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:COLEC7|SFTP4|SP-D|PSP-D
- Protein/Peptide Name:SFTPD
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant rhesus SFTPD consists of 365 amino acids and has a calculated molecular mass of 36.8 kDa.
- Sequence:Met1-Phe375
- Shipping Temperature:Ambient
- Cat. No.:103670-100
A DNA sequence encoding the rhesus SFTPD (NP_001035283.1) (Met 1-Phe 375) precursor was expressed with a C-terminal polyhistidine tag.