Mouse Recombinant CD40/TNFRSF5 Protein (Fc Tag) (from HEK293 Cells)
Catalog # 103886-446
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein Function:Immunotherapy
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized mCD40-Fc (Cat:50324-M02H) at 10 μg/ml (100 μl/well) can bind cynoCD40LG-Fc/Biotin (Cat:90096-C01H).
- Gene ID:NP_035741.2
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:TRAP|AI326936|p50|IMD3|GP39|Tnfrsf5|IGM|T-BAM|Bp50|HIGM1
- Protein/Peptide Name:CD40
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant mouse CD40 consists of 415 amino acids and predicts a molecular mass of 46.4 kDa.
- Sequence:Met1-Arg193
- Shipping Temperature:Ambient
- Cat. No.:103886-446
Specifications
About this item
A DNA sequence encoding the mouse CD40 (NP_035741.2) (Met1-Arg193) was expressed with the Fc region of human IgG1 at the C-terminus.