Influenza A H1N2 (A/Swine/Spain/SF12091/2007) Recombinant Neuraminidase/NA Protein (His Tag) (from HEK293 Cells)
Catalog # 103887-490
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Influenza A
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Gene ID:CCQ71972
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Neuraminidase
- Protein/Peptide Name:NA
- Purity:>85% as determined by SDS-PAGE.
- Molecular Weight:The recombinant Influenza virus A (A/Swine/Spain/SF12091/2007 (H1N2)) neuraminidase consists of 453 amino acids and predicts a molecular mass of 50.6 kDa.
- Sequence:His36-Ile469
- Shipping Temperature:Ambient
- Cat. No.:103887-490
Specifications
About this item
A DNA sequence encoding the influenza A virus (A/Swine/Spain/SF12091/2007 (H1N2)) neuraminidase (CCQ71972) (His36-Ile469), termed as NA, was fused with a N-terminal polyhistidine tag.