SARS-CoV (strain WH20) Recombinant Plpro/papain-like protease Protein (His Tag) (from E. coli)
103664-308
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Gene ID:AAX16193.1
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:PLpro
- Protein/Peptide Name:PLpro
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:The recombinant SARS-CoV (strain WH20) papain-like protease (PLpro) consists of 325 amino acids and predicts a molecular mass of 36.65 kDa.
- Sequence:Glu1541-Tyr1859
- Shipping Temperature:Ambient
- Cat. No.:103664-308
A DNA sequence encoding the SARS-CoV (strain WH20) papain-like protease (PLpro) (AAX16193.1) (Glu1541-Tyr1859) was expressed with a polyhistidine tag at the C-terminus.