Influenza A H11N2 (A/thick-billed murre/Newfoundland/031/2007) Recombinant Hemagglutinin/HA Protein (ECD, His Tag) (from Baculovirus Insect Cells)
Catalog # 103663-922
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Influenza A
- Size:100 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method.
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:Hemagglutinin
- Protein/Peptide Name:HA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:The recombinant hemagglutinin of Influenza A virus (A/thick-billed murre/Newfoundland/031/2007(H11N2)) consists 523 amino acids and predicts a molecular mass of 58.7 kDa.
- Sequence:Met1-Lys528
- Shipping Temperature:Ambient
- Cat. No.:103663-922
Specifications
About this item
A DNA sequence encoding the Influenza A virus (A/thick-billed murre/Newfoundland/031/2007(H11N2)) hemagglutinin (ADD21478.1) (Met1-Lys528), termed as HA, was expressed with a polyhistidine tag at the C-terminus.